2-Fluoro-5-iodotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Fluoro-4-iodo-2-methylbenzene, 2-Fluoro-5-iodo-1-methylbenzene

005313-1G 005313-5G 005313-25G
FLUO005313-1GEA 0 GBP
FLUO005313-1G FLUO005313-5G FLUO005313-25G
2-Fluoro-5-iodotoluene
2-Fluoro-5-iodotoluene
Formula: C₇H₆FI
MW: 236.03 g/mol
MDL Number: MFCD00013710
CAS Number: 452-68-6
EINECS: 207-206-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR