4-Fluoro-2-iodotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-Fluoro-3-iodo-4-methylbenzene, 4-Fluoro-2-iodo-1-methylbenzene
006249-5G
006249-25G
FLUO006249-5GEA
0
GBP
FLUO006249-5G
FLUO006249-25G
4-Fluoro-2-iodotoluene
4-Fluoro-2-iodotoluene
Formula:
C₇H₆FI MW: 236.03 g/mol |
MDL Number:
MFCD00013709 CAS Number: 13194-67-7 EINECS: 236-153-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...