α,α,α,3-Tetrafluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Fluorobenzotrifluoride, 1-Fluoro-3-(trifluoromethyl)benzene
001421-25G
001421-100G
001421-500G
FLUO001421-25GEA
0
GBP
FLUO001421-25G
FLUO001421-100G
FLUO001421-500G
α,α,α,3-Tetrafluorotoluene
α,α,α,3-Tetrafluorotoluene
Formula:
C₇H₄F₄ MW: 164.1 g/mol Boiling Pt: 100 °C (1013 hPa) Melting Pt: –81.5 °C Density: 1.3021 g/cm³ (16.9 °C) |
MDL Number:
MFCD00000382 CAS Number: 401-80-9 EINECS: 206-933-1 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...