2,6-Difluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Difluoro-2-methylbenzene

003205-25G 003205-5G
FLUO003205-25GEA 0 GBP
FLUO003205-25G FLUO003205-5G
2,6-Difluorotoluene
2,6-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 111…112 °C (1013 hPa)
Density: 1.1296 g/cm³ (20 °C)
MDL Number: MFCD00043898
CAS Number: 443-84-5
UN: 2388
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR