4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Chloro-2,6-dinitro-4-(trifluoromethyl)benzene, 4-Chloro-3,5-dinitrobenzotrifluoride
002687-25G
002687-100G
FLUO002687-25GEA
0
GBP
FLUO002687-25G
FLUO002687-100G
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Formula:
C₇H₂ClF₃N₂O₄ MW: 270.55 g/mol Boiling Pt: 250 °C (1013 hPa) Melting Pt: 56…58 °C Density: 1.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007068 CAS Number: 393-75-9 EINECS: 206-889-3 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...