4-Chloro-2-fluoro-5-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010675-5G 010675-25G
FLUO010675-5GEA 0 GBP
FLUO010675-5G FLUO010675-25G
4-Chloro-2-fluoro-5-nitrotoluene
4-Chloro-2-fluoro-5-nitrotoluene
Formula: C₇H₅ClFNO₂
MW: 189.57 g/mol
MDL Number: MFCD00134231
CAS Number: 18349-11-6
EINECS: 242-225-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR