4-Chloro-3-nitrobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Chloro-2-nitro-4-(trifluoromethyl)benzene, 4-Chloro-α-α-α-trifluoro-3-nitrotoluene, 4-Chloro-3-nitrobenzotrifluoride
004508-250G
004508-1KG
FLUO004508-250GEA
0
GBP
FLUO004508-250G
FLUO004508-1KG
4-Chloro-3-nitrobenzotrifluoride
4-Chloro-3-nitrobenzotrifluoride
Formula:
C₇H₃ClF₃NO₂ MW: 225.55 g/mol Boiling Pt: 223 °C (1013 hPa) Melting Pt: –3 °C Density: 1.535 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007084 CAS Number: 121-17-5 EINECS: 204-451-6 UN: 2307 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...