1-(2-Chloroethyl)-4-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

010403-1G 010403-5G
FLUO010403-1GEA 0 GBP
FLUO010403-1G FLUO010403-5G
1-(2-Chloroethyl)-4-fluorobenzene
1-(2-Chloroethyl)-4-fluorobenzene
Formula: C₈H₈ClF
MW: 158.6 g/mol
MDL Number: MFCD00155000
CAS Number: 332-43-4
EINECS: 206-364-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR