2,3-Dichlorotoluene

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

EHERC12505600EA 89.7 GBP
EHERC12505600
2,3-Dichlorotoluene
2,3-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.228 g/cm³ (20 °C)
MDL Number: MFCD00000546
CAS Number: 32768-54-0
EINECS: 251-203-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR