2,4-Difluorobenzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene

001922-5G 001922-25G 001922-100G
FLUO001922-5GEA 0 GBP
FLUO001922-5G FLUO001922-25G FLUO001922-100G
2,4-Difluorobenzyl bromide
2,4-Difluorobenzyl bromide
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 42 °C (1 mmHg)
Density: 1.61 g/cm³ (20 °C)
MDL Number: MFCD00011649
CAS Number: 23915-07-3
EINECS: 245-938-3
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR