Pentafluorobenzyl bromide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001353-25G
001353-100G
001353-10G
FLUO001353-25GEA
0
GBP
FLUO001353-25G
FLUO001353-100G
FLUO001353-10G
Pentafluorobenzyl bromide
Pentafluorobenzyl bromide
Formula:
(C₆F₅)CH₂Br MW: 260.99 g/mol Boiling Pt: 174 °C (1013 hPa) Density: 1.72 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000299 CAS Number: 1765-40-8 EINECS: 217-182-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...