5-Bromo-2-chlorobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-4-chloro-3-(trifluoromethyl)benzene, 5-Bromo-2-chlorobenzotrifluoride, 4-Bromo-1-chloro-2-(trifluoromethyl)benzene, 5-Bromo-2-chloro-α,α,α-trifluorotoluene

004147-25G 004147-100G 004147-500G
FLUO004147-25GEA 0 GBP
FLUO004147-25G FLUO004147-100G FLUO004147-500G
5-Bromo-2-chlorobenzotrifluoride
5-Bromo-2-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –1,4 °C
Density: 1,7486 g/cm³ (25 °C)
MDL Number: MFCD00000601
CAS Number: 445-01-2
EINECS: 207-149-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR