1-Benzyloxy-2-bromobenzene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Bromo-2-benzyloxybenzene, 1-(Benzyloxy)-2-bromobenzene
M-Clarity™ quality level = MQ100
519642-25G
519642-5G
SIAL519642-25GEA
546
GBP
SIAL519642-25G
SIAL519642-5G
1-Benzyloxy-2-bromobenzene, Sigma-Aldrich®
1-Benzyloxy-2-bromobenzene
Formula:
C₁₃H₁₁BrO MW: 263.13 g/mol |
MDL Number:
MFCD03427006 CAS Number: 31575-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...