(3-Chloro-2,6-difluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005607-1G 005607-5G
FLUO005607-1GEA 0 GBP
FLUO005607-1G FLUO005607-5G
(3-Chloro-2,6-difluorophenyl)acetic acid
(3-Chloro-2,6-difluorophenyl)acetic acid
Formula: C₈H₅ClF₂O₂
MW: 206.58 g/mol
MDL Number: MFCD01631328
CAS Number: 261762-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR