5-Fluoro-3-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoropyridine-5-boronic acid pinacol ester, 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-3512-1G PN-3512-5G
COBBPN-3512-1GEA 0 GBP
COBBPN-3512-1G COBBPN-3512-5G
5-Fluoro-3-pyridineboronic acid pinacol ester
5-Fluoro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD07780754
CAS Number: 719268-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR