2-Phenylethylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Phenethylboronic Acid Pinacol Ester, 2-Phenylethyl-1-boronic acid pinacol ester

PN-3483-5G PN-3483-1G
COBBPN-3483-5GEA 0 GBP
COBBPN-3483-5G COBBPN-3483-1G
2-Phenylethylboronic acid pinacol ester
2-Phenylethylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₂
MW: 232.13 g/mol
MDL Number: MFCD03788721
CAS Number: 165904-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR