2,4-Dimethoxyphenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(2,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2667-5G PN-2667-1G
COBBPN-2667-5GEA 0 GBP
COBBPN-2667-5G COBBPN-2667-1G
2,4-Dimethoxyphenylboronic acid pinacol ester
2,4-Dimethoxyphenylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₄
MW: 264.13 g/mol
MDL Number: MFCD06795688
CAS Number: 214360-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR