2,6-Difluoro-3-isopropoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2,6-Difluoro-3-isopropoxyphenyl)boronic acid

BB-8065-1G BB-8065-250M
COBBBB-8065-1GEA 0 GBP
COBBBB-8065-1G COBBBB-8065-250M
2,6-Difluoro-3-isopropoxyphenylboronic acid
2,6-Difluoro-3-isopropoxyphenylboronic acid
Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
MDL Number: MFCD07369753
CAS Number: 849062-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR