2-Bromo-4-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Bromo-4-fluorophenyl)boronic acid

BB-5964-1G BB-5964-5G BB-5964-250M
COBBBB-5964-1GEA 0 GBP
COBBBB-5964-1G COBBBB-5964-5G COBBBB-5964-250M
2-Bromo-4-fluorophenylboronic acid
2-Bromo-4-fluorophenylboronic acid
Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
MDL Number: MFCD13195647
CAS Number: 1217501-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR