2,3,4-Trichlorophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5217-5G
BB-5217-1G
COBBBB-5217-5GEA
0
GBP
COBBBB-5217-5G
COBBBB-5217-1G
2,3,4-Trichlorophenylboronic acid
2,3,4-Trichlorophenylboronic acid
Formula:
C₆H₄BCl₃O₂ |
MDL Number:
MFCD03095112 CAS Number: 352530-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...