2,6-Difluoro-4-methoxyphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5042-5G
BB-5042-25G
BB-5042-1G
COBBBB-5042-5GEA
0
GBP
COBBBB-5042-5G
COBBBB-5042-25G
COBBBB-5042-1G
2,6-Difluoro-4-methoxyphenylboronic acid
2,6-Difluoro-4-methoxyphenylboronic acid
Formula:
C₇H₇BF₂O₃ MW: 187.94 g/mol |
MDL Number:
MFCD05664299 CAS Number: 406482-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...