2,3-Difluoro-5-nitrophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2,3-Difluoro-5-nitrophenyl)boronic acid

BB-3801-1G BB-3801-5G
COBBBB-3801-1GEA 0 GBP
COBBBB-3801-1G COBBBB-3801-5G
2,3-Difluoro-5-nitrophenylboronic acid
2,3-Difluoro-5-nitrophenylboronic acid
Formula: C₆H₄BF₂NO₄
MW: 202.91 g/mol
MDL Number: MFCD09027242
CAS Number: 957060-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR