2,4-Difluoro-5-nitrophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3589-5G BB-3589-1G
COBBBB-3589-5GEA 0 GBP
COBBBB-3589-5G COBBBB-3589-1G
2,4-Difluoro-5-nitrophenylboronic acid
2,4-Difluoro-5-nitrophenylboronic acid
Formula: C₆H₄BF₂NO₄
MW: 202.91 g/mol
MDL Number: MFCD03095253
CAS Number: 325786-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR