2-Bromo-6-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Bromo-6-fluorophenyl)boronic acid, 2-Bromo-6-fluorobenzeneboronic acid

BB-3498-25G BB-3498-5G
COBBBB-3498-25GEA 0 GBP
COBBBB-3498-25G COBBBB-3498-5G
2-Bromo-6-fluorophenylboronic acid
2-Bromo-6-fluorophenylboronic acid
Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
MDL Number: MFCD05664309
CAS Number: 913835-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR