2,6-Difluoro-3-nitrophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,6-Difluoro-3-nitrobenzeneboronic acid, (2,6-Difluoro-3-nitrophenyl)boronic acid
BB-2144-5G
BB-2144-1G
COBBBB-2144-5GEA
0
GBP
COBBBB-2144-5G
COBBBB-2144-1G
2,6-Difluoro-3-nitrophenylboronic acid
2,6-Difluoro-3-nitrophenylboronic acid
Formula:
C₆H₄BF₂NO₄ MW: 202.91 g/mol |
MDL Number:
MFCD12025957 CAS Number: 1150114-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...