Allylbenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Phenyl-2-propene, 3-Phenylpropene

005425-25G 005425-100G
FLUO005425-25GEA 0 GBP
FLUO005425-25G FLUO005425-100G
Allylbenzene
Allylbenzene
Formula: (C₆H₅)CH₂CHCH₂
MW: 118.18 g/mol
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: < –40 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00008651
CAS Number: 300-57-2
EINECS: 206-095-7
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR