Allylpentafluorobenzene ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-(Pentafluorophenyl)-1-propene, 1-Allyl-2,3,4,5,6-pentafluorobenzene
A17834.06
A17834.14
A17834.06EA
30.9
GBP
A17834.06
A17834.14
Allylpentafluorobenzene ≥98%
Allylpentafluorobenzene
Purity: 98%
CAS No.: 1736-60-3
Molecular Formula: C9H5F5
Formula Weight: 208.13
Flash Point: 42°(108°F)
Refractive Index: 1.4270
Boiling Point: 148-149°
Beilstein Reference: 1971209
UN No.: 1993
MDL No.: MFCD00000301
Formula:
(C₆F₅)CH₂CHCH₂ MW: 208.13 g/mol Boiling Pt: 148…149 °C Density: 1.358 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000301 CAS Number: 1736-60-3 EINECS: 217-083-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...