4'-tert-Butylbiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4875-5G CA-4875-1G
COBBCA-4875-5GEA 0 GBP
COBBCA-4875-5G COBBCA-4875-1G
4'-tert-Butylbiphenyl-3-carboxylic acid
4'-tert-Butylbiphenyl-3-carboxylic acid
Formula: C₁₇H₁₈O₂
MDL Number: MFCD06802635
CAS Number: 81770-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR