3',5'-Dimethylbiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4854-5G CA-4854-1G
COBBCA-4854-5GEA 0 GBP
COBBCA-4854-5G COBBCA-4854-1G
3',5'-Dimethylbiphenyl-3-carboxylic acid
3',5'-Dimethylbiphenyl-3-carboxylic acid
Formula: C₁₅H₁₄O₂
MDL Number: MFCD06201437
CAS Number: 177734-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR