3',4'-Dimethylbiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4896-1G CA-4896-250M
COBBCA-4896-1GEA 0 GBP
COBBCA-4896-1G COBBCA-4896-250M
3',4'-Dimethylbiphenyl-3-carboxylic acid
3',4'-Dimethylbiphenyl-3-carboxylic acid
Formula: C₁₅H₁₄O₂
MDL Number: MFCD04117430
CAS Number: 728919-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR