2-Methyl-4-nitrophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-5486-1G PN-5486-250M
COBBPN-5486-1GEA 0 GBP
COBBPN-5486-1G COBBPN-5486-250M
2-Methyl-4-nitrophenylboronic acid pinacol ester
2-Methyl-4-nitrophenylboronic acid pinacol ester
Formula: C₁₃H₁₈BNO₄
MDL Number: MFCD10698807
CAS Number: 883715-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR