3-Iodophenylboronic acid pinacol ester, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Iodobenzeneboronic acid pinacol ester, 2-(3-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
M-Clarity™ quality level = MQ100
699462-1G
699462-5G
SIAL699462-1GEA
92.9
GBP
SIAL699462-1G
SIAL699462-5G
3-Iodophenylboronic acid pinacol ester, Sigma-Aldrich®
3-Iodophenylboronic acid pinacol ester
Formula:
C₁₂H₁₆BIO₂ MW: 329.97 g/mol |
MDL Number:
MFCD09953498 CAS Number: 408492-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...