3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(3,5-Difluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8716-1G PN-8716-250M
COBBPN-8716-1GEA 0 GBP
COBBPN-8716-1G COBBPN-8716-250M
3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester
3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
MDL Number: MFCD07781207
CAS Number: 1073354-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR