4-Amino-3-fluorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester

PN-8771-5G PN-8771-1G
COBBPN-8771-5GEA 0 GBP
COBBPN-8771-5G COBBPN-8771-1G
4-Amino-3-fluorophenylboronic acid pinacol ester
4-Amino-3-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
MDL Number: MFCD09033884
CAS Number: 819058-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR