4-Chlorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

PN-2261-5G PN-2261-25G PN-2261-1G
COBBPN-2261-5GEA 0 GBP
COBBPN-2261-5G COBBPN-2261-25G COBBPN-2261-1G
4-Chlorophenylboronic acid pinacol ester
4-Chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BClO₂
MW: 238.52 g/mol
MDL Number: MFCD05663875
CAS Number: 195062-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR