3,5-Dichlorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(3,5-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2269-5G PN-2269-1G
COBBPN-2269-5GEA 0 GBP
COBBPN-2269-5G COBBPN-2269-1G
3,5-Dichlorophenylboronic acid pinacol ester
3,5-Dichlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BCl₂O₂
MW: 272.97 g/mol
MDL Number: MFCD05863906
CAS Number: 68716-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR