2-Nitrophenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Nitrophenylboronic acid Pinacol ester, 2-Nitrobenzeneboronic acid pinacol ester, 2-Nitrophenylboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane, 1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
BB-2120-1G
BB-2120-5G
COBBBB-2120-1GEA
0
GBP
COBBBB-2120-1G
COBBBB-2120-5G
2-Nitrophenylboronic acid pinacol ester
2-Nitrophenylboronic acid pinacol ester
Formula:
C₁₂H₁₆BNO₄ MW: 249.07 g/mol |
MDL Number:
MFCD02179447 CAS Number: 190788-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...