3-Cyanophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

PN-2455-5G PN-2455-1G
COBBPN-2455-5GEA 0 GBP
COBBPN-2455-5G COBBPN-2455-1G
3-Cyanophenylboronic acid pinacol ester
3-Cyanophenylboronic acid pinacol ester
Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
MDL Number: MFCD03789262
CAS Number: 214360-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR