1-Pentenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Penten-1-ylboronic acid

M-Clarity™ quality level = MQ100

578452-1G 578452-5G
SIAL578452-1GEA 162 GBP
SIAL578452-1G SIAL578452-5G
1-Pentenylboronic acid, Sigma-Aldrich®
1-Pentenylboronic acid
Formula: C₅H₁₁BO₂
MW: 113.95 g/mol
MDL Number: MFCD01074677
CAS Number: 104376-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR