Ethyl isothiocyanatoformate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethoxycarbonyl isothiocyanate

E1473.10 E1473.50
XXXXE1473.10EA 0 GBP
XXXXE1473.10 XXXXE1473.50
Ethyl isothiocyanatoformate
Ethyl isothiocyanatoformate
Formula: C₄H₅NO₂S
MW: 131.16 g/mol
MDL Number: MFCD00004814
CAS Number: 16182-04-0
EINECS: 240-318-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR