1-Cyanocyclopropanecarboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Cyano-1-cyclopropanecarboxylic acid

001453-10G 001453-1G 001453-25G 001453-5G
FLUO001453-10GEA 0 GBP
FLUO001453-10G FLUO001453-1G FLUO001453-25G FLUO001453-5G
1-Cyanocyclopropanecarboxylic acid
1-Cyanocyclopropanecarboxylic acid
Formula: C₅H₅NO₂
MW: 111.1 g/mol
MDL Number: MFCD00190651
CAS Number: 6914-79-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR