4'-Bromobiphenyl-4-carboxylic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4'-Bromo[1,1'-biphenyl]-4-carboxylic acid, 4'-Bromo-[1,1'-biphenyl]-4-carboxylic acid

3T-0295.500 3T-0295.1 3T-0295.5
BIOT3T-0295.500EA 0 GBP
BIOT3T-0295.500 BIOT3T-0295.1 BIOT3T-0295.5
4'-Bromobiphenyl-4-carboxylic acid
4'-Bromobiphenyl-4-carboxylic acid
Formula: C₁₃H₉BrO₂
MW: 277.12 g/mol
MDL Number: MFCD00452920
CAS Number: 5731-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR