4-Bromo-3-(trifluoromethyl)benzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7560-5G OS-7560-25G OS-7560-1G
COBBOS-7560-5GEA 0 GBP
COBBOS-7560-5G COBBOS-7560-25G COBBOS-7560-1G
4-Bromo-3-(trifluoromethyl)benzonitrile
4-Bromo-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MDL Number: MFCD03095347
CAS Number: 1735-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR