2-Bromo-5-(trifluoromethyl)benzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4407-25G CA-4407-5G CA-4407-100G
COBBCA-4407-25GEA 0 GBP
COBBCA-4407-25G COBBCA-4407-5G COBBCA-4407-100G
2-Bromo-5-(trifluoromethyl)benzonitrile
2-Bromo-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MDL Number: MFCD03412187
CAS Number: 1483-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR