2-Fluorobenzothiazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006191-1G
006191-5G
FLUO006191-1GEA
0
GBP
FLUO006191-1G
FLUO006191-5G
2-Fluorobenzothiazole
2-Fluorobenzothiazole
Formula:
C₇H₄FNS MW: 153.18 g/mol |
MDL Number:
MFCD00047254 CAS Number: 1123-98-4 EINECS: 214-384-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...