1-Boc-6-methylindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0186-5G IN-0186-1G
COBBIN-0186-5GEA 0 GBP
COBBIN-0186-5G COBBIN-0186-1G
1-Boc-6-methylindole
1-Boc-6-methylindole
Formula: C₁₄H₁₇NO₂
MW: 231.29 g/mol
MDL Number: MFCD11504944
CAS Number: 127956-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR