1-Boc-6-methoxyindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0153-1G IN-0153-5G
COBBIN-0153-1GEA 0 GBP
COBBIN-0153-1G COBBIN-0153-5G
1-Boc-6-methoxyindole
1-Boc-6-methoxyindole
Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD08272247
CAS Number: 138344-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR