Methyl indole-5-carboxylate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 1H-indole-5-carboxylate

MOLEM41072002EA 0 GBP
MOLEM41072002
Methyl indole-5-carboxylate
Methyl indole-5-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00153023
CAS Number: 1011-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR