Methyl indole-4-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IN-0001-25G
IN-0001-5G
COBBIN-0001-25GEA
0
GBP
COBBIN-0001-25G
COBBIN-0001-5G
Methyl indole-4-carboxylate
Methyl indole-4-carboxylate
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol |
MDL Number:
MFCD00191222 CAS Number: 39830-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...