3-Acetoxyindole ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Indolyl acetate3-Indolyl Acetate3-Indoxyl acetateIndoxyl acetate

L04932.03EA 37.9 GBP
L04932.03
3-Acetoxyindole ≥97%
3-Acetoxyindole
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00014561
CAS Number: 608-08-2
EINECS: 210-154-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR